Assay Details
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012893.1 | NP_037025.1 | 3 - 4 | 375 | 68 | |
XM_006236728.2 | XP_006236790.1 | 4 - 5 | 496 | 68 | ||
GenBank mRNA | BC087689.1 | - | 4 - 5 | 441 | 68 | |
FQ220056.1 | - | 4 - 5 | 440 | 68 | ||
FQ220823.1 | - | 4 - 5 | 441 | 68 | ||
FQ221892.1 | - | 4 - 5 | 440 | 68 | ||
FQ223160.1 | - | 4 - 5 | 466 | 68 | ||
FQ229904.1 | - | 4 - 5 | 441 | 68 | ||
M22323.1 | - | 3 - 4 | 375 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
