Assay Details
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243174.2 | XP_006243236.1 | 1 - 2 | 252 | 84 | |
XM_008766330.1 | XP_008764552.1 | 1 - 2 | 252 | 84 | ||
GenBank mRNA | BC070947.1 | - | 1 - 2 | 108 | 84 | |
X99337.1 | - | 1 - 2 | 281 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
