Assay Details
Gene Symbol: Tmprss13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127528.1 | NP_001121000.1 | 10 - 11 | 1314 | 83 | |
GenBank mRNA | BC166801.1 | - | 10 - 11 | 1314 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
