Assay Details
Gene Symbol: Pafah1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022387.3 | NP_071782.1 | 4 - 5 | 408 | 68 | |
XM_006242957.2 | XP_006243019.1 | 4 - 5 | 874 | 68 | ||
XM_006242958.2 | XP_006243020.1 | 4 - 5 | 407 | 68 | ||
XM_008766172.1 | XP_008764394.1 | 4 - 5 | 638 | 68 | ||
GenBank mRNA | AF016048.1 | - | 4 - 5 | 371 | 68 | |
BC081714.1 | - | 4 - 5 | 408 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
