Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013168.2 | NP_037300.2 | 10 - 11 | 785 | 111 | |
XM_006242896.1 | XP_006242958.1 | 10 - 11 | 871 | 111 | ||
XM_006242897.1 | XP_006242959.1 | 9 - 10 | 793 | 111 | ||
XM_008766148.1 | XP_008764370.1 | 9 - 10 | 955 | 111 | ||
XM_008766149.1 | XP_008764371.1 | 9 - 10 | 892 | 111 | ||
GenBank mRNA | BC088162.1 | - | 10 - 11 | 785 | 111 | |
FQ225972.1 | - | 4 - 5 | 270 | 111 | ||
FQ226062.1 | - | 10 - 11 | 708 | 111 | ||
FQ234793.1 | - | 2 - 3 | 157 | 111 | ||
FQ234870.1 | - | 8 - 9 | 941 | 111 | ||
X06827.1 | - | 10 - 11 | 660 | 111 | ||
Y12006.1 | - | 10 - 11 | 655 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
