Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013168.2 | NP_037300.2 | 7 - 8 | 557 | 139 | |
XM_006242896.1 | XP_006242958.1 | 7 - 8 | 643 | 139 | ||
XM_006242897.1 | XP_006242959.1 | 6 - 7 | 565 | 139 | ||
XM_008766148.1 | XP_008764370.1 | 6 - 7 | 727 | 139 | ||
XM_008766149.1 | XP_008764371.1 | 6 - 7 | 664 | 139 | ||
GenBank mRNA | BC088162.1 | - | 7 - 8 | 557 | 139 | |
FQ226062.1 | - | 7 - 8 | 480 | 139 | ||
FQ231091.1 | - | 7 - 8 | 541 | 139 | ||
FQ234870.1 | - | 5 - 6 | 713 | 139 | ||
X06827.1 | - | 7 - 8 | 432 | 139 | ||
Y12006.1 | - | 7 - 8 | 427 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
