Assay Details
Gene Symbol: Cib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015010.1 | NP_001015010.1 | 2 - 3 | 258 | 66 | |
GenBank mRNA | BC092630.1 | - | 2 - 3 | 258 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
