Assay Details
Gene Symbol: Cib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015010.1 | NP_001015010.1 | 4 - 5 | 513 | 122 | |
XM_006243071.2 | XP_006243133.1 | 3 - 4 | 456 | 122 | ||
GenBank mRNA | BC092630.1 | - | 4 - 5 | 513 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
