Assay Details
Gene Symbol: Dpagt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199388.1 | NP_955420.1 | 3 - 4 | 632 | 74 | |
XM_006242921.2 | XP_006242983.1 | 3 - 4 | 983 | 74 | ||
GenBank mRNA | BC063184.1 | - | 3 - 4 | 632 | 74 | |
FQ209511.1 | - | 1 - 2 | 71 | 74 | ||
FQ213783.1 | - | 3 - 4 | 709 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
