Assay Details
Gene Symbol: Dpagt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199388.1 | NP_955420.1 | 7 - 8 | 1136 | 135 | |
GenBank mRNA | BC063184.1 | - | 7 - 8 | 1136 | 135 | |
FQ209511.1 | - | 5 - 6 | 575 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
