Assay Details
Gene Symbol: Rbpms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173426.1 | NP_001166897.1 | 8 - 9 | 789 | 69 | |
XM_006243274.2 | XP_006243336.1 | 8 - 9 | 2475 | 69 | ||
XM_008766329.1 | XP_008764551.1 | 7 - 8 | 898 | 69 | ||
GenBank mRNA | BC169038.1 | - | 8 - 9 | 789 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
