Assay Details
Gene Symbol: Cyp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012541.3 | NP_036673.3 | 5 - 6 | 1211 | 136 | |
GenBank mRNA | BC127476.1 | - | 5 - 6 | 1211 | 136 | |
FQ209607.1 | - | 4 - 5 | 639 | 136 | ||
FQ210213.1 | - | 5 - 6 | 1217 | 136 | ||
FQ210219.1 | - | 4 - 5 | 359 | 136 | ||
K02422.1 | - | 4 - 5 | 1179 | 136 | ||
M26127.1 | - | 4 - 5 | 368 | 136 | ||
X01031.1 | - | 4 - 5 | 726 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
