Assay Details
Gene Symbol: Cyp19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017085.2 | NP_058781.2 | 1 - 2 | 342 | 120 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_017085.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
