Assay Details
Gene Symbol: Fsd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763722.1 | XP_008761944.1 | 1 - 2 | 199 | 81 | |
XM_008763723.1 | XP_008761945.1 | 1 - 2 | 198 | 81 | ||
XM_008763724.1 | XP_008761946.1 | 1 - 2 | 199 | 81 | ||
XM_008763725.1 | XP_008761947.1 | 1 - 2 | 200 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008763722.1, XM_008763723.1, XM_008763724.1, XM_008763725.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
