Assay Details
Gene Symbol: Serinc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008312.1 | NP_001008313.1 | 1 - 2 | 133 | 112 | |
GenBank mRNA | BC085853.1 | - | 1 - 2 | 133 | 112 | |
EU220431.1 | - | 1 - 2 | 79 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
