Assay Details
Gene Symbol: Serinc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008312.1 | NP_001008313.1 | 8 - 9 | 1145 | 169 | |
GenBank mRNA | BC085853.1 | - | 8 - 9 | 1145 | 169 | |
EU220431.1 | - | 8 - 9 | 1091 | 169 | ||
FQ222836.1 | - | 2 - 3 | 228 | 169 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
