Assay Details
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201635.2 | NP_963966.1 | 1 - 2 | 260 | 80 | |
XM_006235672.2 | XP_006235734.1 | 1 - 2 | 602 | 80 | ||
GenBank mRNA | BC100059.1 | - | 1 - 2 | 605 | 80 | |
X59506.1 | - | 1 - 2 | 603 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
