Assay Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073089.1 | 2 - 3 | 286 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
