Assay Details
Gene Symbol: Cdh22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019161.2 | NP_062034.2 | 11 - 12 | 2307 | 61 | |
GenBank mRNA | D83348.1 | - | 11 - 12 | 2313 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P033979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50867 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P033979 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P033979 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P033979 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P454259 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P116532 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62319 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57993 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20849 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99295 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40053 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102385 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38961 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95426 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10851726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851734 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851725 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851730 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851731 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10851724 | - | - |
Rat Genome 230 2.0 Array | 1388045_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1388045_a_at | - | - |
Rat Genome 230 2.0 Array | 1368887_at | - | - |
Rat Genome 230 2.0 Array | 1368887_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388045_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368887_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368887_at | - | - |
Affymetrix HT Rat Focus Array | 1388045_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300036546.rn.1 | TC0300004770.rn.1 | RTA TC |
Rat Expression Set 230 | 1368887_PM_at | - | - |
Rat Expression Set 230 | 1368887_at | - | - |
Rat Expression Set 230 | 1388045_PM_a_at | - | - |
Rat Expression Set 230 | 1388045_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851724 | - | - |
Rat Genome U34 Set | D83348_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21633870 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
RatRef-12 Expression BeadChip | ILMN_50959 | - | - |
