Assay Details
Gene Symbol: Cdh22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019161.2 | NP_062034.2 | 4 - 5 | 1063 | 87 | |
GenBank mRNA | D83348.1 | - | 4 - 5 | 1069 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
