Assay Details
Gene Symbol: Srsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014185.2 | NP_001014207.2 | 5 - 6 | 801 | 73 | |
GenBank mRNA | BC087121.1 | - | 5 - 6 | 792 | 73 | |
FM081417.1 | - | 3 - 4 | 325 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
