Assay Details
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235520.2 | XP_006235582.1 | 2 - 3 | 136 | 75 | |
XM_006235521.1 | XP_006235583.1 | 2 - 3 | 228 | 75 | ||
XM_008762469.1 | XP_008760691.1 | 1 - 2 | 1506 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235520.2, XM_006235521.1, XM_008762469.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
