Assay Details
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024381.2 | NP_077357.2 | 17 - 18 | 1579 | 60 | |
XM_006256987.2 | XP_006257049.1 | 18 - 19 | 1631 | 60 | ||
XM_006256988.2 | XP_006257050.1 | 17 - 18 | 1613 | 60 | ||
XM_008773274.1 | XP_008771496.1 | 19 - 20 | 1696 | 60 | ||
XM_008773275.1 | XP_008771497.1 | 18 - 19 | 1678 | 60 | ||
XM_008773276.1 | XP_008771498.1 | 19 - 20 | 1696 | 60 | ||
XM_008773278.1 | XP_008771500.1 | 19 - 20 | 1695 | 60 | ||
GenBank mRNA | CO557097.1 | - | 2 - 3 | 261 | 60 | |
D16102.1 | - | 17 - 18 | 1582 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
