Assay Details
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024381.2 | NP_077357.2 | 18 - 19 | 1657 | 109 | |
XM_008773278.1 | XP_008771500.1 | 20 - 21 | 1773 | 109 | ||
GenBank mRNA | CO557097.1 | - | 3 - 4 | 339 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
glycerol metabolic process
glycerol-3-phosphate metabolic process
triglyceride metabolic process
response to cold
response to xenobiotic stimulus
response to organic substance
phosphorylation
glycerol catabolic process
glucose homeostasis
response to ethanol
glycerol-3-phosphate biosynthetic process
glycerol kinase activity
ATP binding
histone binding
glycerol metabolic process
glycerol-3-phosphate metabolic process
triglyceride metabolic process
response to cold
response to xenobiotic stimulus
response to organic substance
phosphorylation
glycerol catabolic process
glucose homeostasis
response to ethanol
glycerol-3-phosphate biosynthetic process
glycerol kinase activity
ATP binding
histone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P123949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170453 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123949 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123949 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P123949 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P148071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104498 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4460 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23927 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39541 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60146 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10938234 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938242 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938232 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938241 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938240 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938231 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938235 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938233 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938239 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938236 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938220 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938225 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938238 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938229 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10938219 | - | - |
Rat Genome 230 2.0 Array | 1391661_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387491_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391661_at | - | - |
Rat Genome 230 2.0 Array | 1387491_at | - | - |
Affymetrix HT Rat Focus Array | 1387491_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391661_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391661_at | - | - |
Affymetrix HT Rat Focus Array | 1387491_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00011488.rn.1 | TC0X00001797.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011489.rn.1 | TC0X00001797.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011486.rn.1 | TC0X00001797.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011485.rn.1 | TC0X00001797.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011490.rn.1 | TC0X00001797.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011487.rn.1 | TC0X00001797.rn.1 | RTA TC |
Rat Expression Set 230 | 1387491_at | - | - |
Rat Expression Set 230 | 1387491_PM_at | - | - |
Rat Expression Set 230 | 1391661_PM_at | - | - |
Rat Expression Set 230 | 1391661_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938219 | - | - |
Rat Genome U34 Set | D16102_at | - | - |
Rat Genome U34 Set | rc_AA945076_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21089782 | - | - |
