Assay Details
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012502.1 | NP_036634.1 | 2 - 3 | 2702 | 120 | |
GenBank mRNA | M20133.1 | - | 2 - 3 | 2702 | 120 | |
M23264.1 | - | 2 - 3 | 1741 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
