Assay Details
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012502.1 | NP_036634.1 | 3 - 4 | 2821 | 108 | |
GenBank mRNA | M20133.1 | - | 3 - 4 | 2821 | 108 | |
M23264.1 | - | 3 - 4 | 1860 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
