Assay Details
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012502.1 | NP_036634.1 | 7 - 8 | 3542 | 102 | |
GenBank mRNA | M20133.1 | - | 7 - 8 | 3542 | 102 | |
M23264.1 | - | 7 - 8 | 2581 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
regulation of systemic arterial blood pressure
epithelial cell morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
spermatogenesis
single fertilization
positive regulation of cell population proliferation
male gonad development
fertilization
cellular process
regulation of gene expression
male somatic sex determination
androgen receptor signaling pathway
intracellular receptor signaling pathway
response to insulin
positive regulation of intracellular estrogen receptor signaling pathway
Leydig cell differentiation
multicellular organism growth
positive regulation of phosphorylation
steroid hormone mediated signaling pathway
positive regulation of MAPK cascade
positive regulation of insulin-like growth factor receptor signaling pathway
negative regulation of integrin biosynthetic process
positive regulation of integrin biosynthetic process
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
positive regulation of transcription by RNA polymerase III
male sex differentiation
reproductive structure development
regulation of developmental growth
animal organ formation
male genitalia morphogenesis
negative regulation of epithelial cell proliferation
regulation of catalytic activity
positive regulation of NF-kappaB transcription factor activity
protein complex oligomerization
activation of prostate induction by androgen receptor signaling pathway
morphogenesis of an epithelial fold
lateral sprouting involved in mammary gland duct morphogenesis
regulation of prostatic bud formation
prostate gland growth
prostate gland epithelium morphogenesis
epithelial cell differentiation involved in prostate gland development
tertiary branching involved in mammary gland duct morphogenesis
mammary gland alveolus development
reproductive system development
cellular response to steroid hormone stimulus
seminiferous tubule development
regulation of protein localization to plasma membrane
negative regulation of extrinsic apoptotic signaling pathway
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA binding
chromatin binding
DNA-binding transcription factor activity
nuclear receptor activity
signaling receptor binding
androgen binding
protein binding
beta-catenin binding
transcription factor binding
zinc ion binding
enzyme binding
protein domain specific binding
sequence-specific DNA binding
androgen receptor binding
POU domain binding
in utero embryonic development
regulation of systemic arterial blood pressure
epithelial cell morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
spermatogenesis
single fertilization
positive regulation of cell population proliferation
male gonad development
fertilization
cellular process
regulation of gene expression
male somatic sex determination
androgen receptor signaling pathway
intracellular receptor signaling pathway
response to insulin
positive regulation of intracellular estrogen receptor signaling pathway
Leydig cell differentiation
multicellular organism growth
positive regulation of phosphorylation
steroid hormone mediated signaling pathway
positive regulation of MAPK cascade
positive regulation of insulin-like growth factor receptor signaling pathway
negative regulation of integrin biosynthetic process
positive regulation of integrin biosynthetic process
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
positive regulation of transcription by RNA polymerase III
male sex differentiation
reproductive structure development
regulation of developmental growth
animal organ formation
male genitalia morphogenesis
negative regulation of epithelial cell proliferation
regulation of catalytic activity
positive regulation of NF-kappaB transcription factor activity
protein complex oligomerization
activation of prostate induction by androgen receptor signaling pathway
morphogenesis of an epithelial fold
lateral sprouting involved in mammary gland duct morphogenesis
regulation of prostatic bud formation
prostate gland growth
prostate gland epithelium morphogenesis
epithelial cell differentiation involved in prostate gland development
tertiary branching involved in mammary gland duct morphogenesis
mammary gland alveolus development
reproductive system development
cellular response to steroid hormone stimulus
seminiferous tubule development
regulation of protein localization to plasma membrane
negative regulation of extrinsic apoptotic signaling pathway
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA binding
chromatin binding
DNA-binding transcription factor activity
nuclear receptor activity
signaling receptor binding
androgen binding
protein binding
beta-catenin binding
transcription factor binding
zinc ion binding
enzyme binding
protein domain specific binding
sequence-specific DNA binding
androgen receptor binding
POU domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P037631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166201 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037631 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P037631 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P546901 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10764 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68499 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15538 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 758 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8670 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24302 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66938 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38617 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75280 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32992 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78337 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64744 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46985 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10934125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934126 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934127 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934121 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10934118 | - | - |
Rat Genome 230 2.0 Array | 1369159_at | - | - |
Rat Genome 230 2.0 Array | 1369159_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369159_at | - | - |
Affymetrix HT Rat Focus Array | 1369159_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00003517.rn.1 | TC0X00000570.rn.1 | RTA TC |
Rat Expression Set 230 | 1369159_PM_at | - | - |
Rat Expression Set 230 | 1369159_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934118 | - | - |
Rat Genome U34 Set | M23264_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21272180 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353640 | - | - |
RatRef-12 Expression BeadChip | ILMN_68782 | - | - |
