Assay Details
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106581.1 | NP_001100051.1 | 1 - 2 | 116 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106581.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
