Assay Details
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004072.2 | NP_001004072.2 | 3 - 4 | 328 | 56 | |
GenBank mRNA | BC079369.1 | - | 3 - 4 | 320 | 56 | |
BC098897.1 | - | 3 - 4 | 328 | 56 | ||
Z12158.1 | - | 3 - 4 | 347 | 56 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
