Assay Details
Gene Symbol: Dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013097.2 | NP_037229.1 | 6 - 7 | 828 | 101 | |
XM_006245829.2 | XP_006245891.1 | 7 - 8 | 1508 | 101 | ||
GenBank mRNA | AF397150.1 | - | 6 - 7 | 828 | 101 | |
AF397151.1 | - | 7 - 8 | 699 | 101 | ||
BC089764.1 | - | 6 - 7 | 828 | 101 | ||
X56060.1 | - | 6 - 7 | 681 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
