Assay Details
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009619.1 | NP_001009619.1 | 3 - 4 | 284 | 58 | |
GenBank mRNA | BC088221.1 | - | 3 - 4 | 284 | 58 | |
FQ218444.1 | - | 3 - 4 | 297 | 58 | ||
FQ220638.1 | - | 3 - 4 | 297 | 58 | ||
FQ222868.1 | - | 3 - 4 | 301 | 58 | ||
FQ229330.1 | - | 3 - 4 | 300 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
