Assay Details
Gene Symbol: Serbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145086.1 | NP_659554.1 | 1 - 2 | 382 | 103 | |
XM_006236616.1 | XP_006236678.1 | 1 - 2 | 430 | 103 | ||
XM_006236617.1 | XP_006236679.1 | 1 - 2 | 430 | 103 | ||
XM_006236618.1 | XP_006236680.1 | 1 - 2 | 430 | 103 | ||
GenBank mRNA | AF388527.1 | - | 1 - 2 | 388 | 103 | |
BC079337.1 | - | 1 - 2 | 382 | 103 | ||
FQ213573.1 | - | 1 - 2 | 425 | 103 | ||
FQ214321.1 | - | 1 - 2 | 381 | 103 | ||
FQ223736.1 | - | 1 - 2 | 403 | 103 | ||
FQ225988.1 | - | 1 - 2 | 97 | 103 | ||
FQ226374.1 | - | 1 - 2 | 403 | 103 | ||
FQ226420.1 | - | 1 - 2 | 97 | 103 | ||
FQ227636.1 | - | 1 - 2 | 403 | 103 | ||
FQ228026.1 | - | 1 - 2 | 393 | 103 | ||
FQ230135.1 | - | 1 - 2 | 409 | 103 | ||
FQ230192.1 | - | 1 - 2 | 393 | 103 | ||
FQ230382.1 | - | 1 - 2 | 103 | |||
FQ231329.1 | - | 1 - 2 | 97 | 103 | ||
FQ231840.1 | - | 1 - 2 | 97 | 103 | ||
FQ232288.1 | - | 1 - 2 | 403 | 103 | ||
FQ233179.1 | - | 1 - 2 | 403 | 103 | ||
FQ233457.1 | - | 1 - 2 | 409 | 103 | ||
FQ233487.1 | - | 1 - 2 | 97 | 103 | ||
FQ233911.1 | - | 1 - 2 | 403 | 103 | ||
FQ234284.1 | - | 1 - 2 | 377 | 103 | ||
FQ234534.1 | - | 1 - 2 | 404 | 103 | ||
FQ235054.1 | - | 1 - 2 | 97 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
