Assay Details
Gene Symbol: Serbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145086.1 | NP_659554.1 | 3 - 4 | 663 | 87 | |
XM_006236616.1 | XP_006236678.1 | 3 - 4 | 711 | 87 | ||
GenBank mRNA | AF388527.1 | - | 3 - 4 | 669 | 87 | |
BC079337.1 | - | 3 - 4 | 663 | 87 | ||
FQ226420.1 | - | 3 - 4 | 378 | 87 | ||
FQ231329.1 | - | 3 - 4 | 378 | 87 | ||
FQ231840.1 | - | 3 - 4 | 378 | 87 | ||
FQ232894.1 | - | 3 - 4 | 685 | 87 | ||
FQ233179.1 | - | 3 - 4 | 684 | 87 | ||
FQ233457.1 | - | 3 - 4 | 690 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
