Assay Details
Gene Symbol: Snca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019169.2 | NP_062042.1 | 2 - 3 | 283 | 81 | |
XM_006236591.2 | XP_006236653.1 | 2 - 3 | 564 | 81 | ||
GenBank mRNA | AF007758.1 | - | 1 - 2 | 146 | 81 | |
AY550005.1 | - | 1 - 2 | 146 | 81 | ||
AY550006.1 | - | 1 - 2 | 146 | 81 | ||
BC087682.1 | - | 2 - 3 | 283 | 81 | ||
FQ212580.1 | - | 2 - 3 | 260 | 81 | ||
S73007.1 | - | 2 - 3 | 214 | 81 | ||
S73008.1 | - | 2 - 3 | 214 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
