Assay Details
Gene Symbol: Mgll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138502.2 | NP_612511.1 | 6 - 7 | 686 | 113 | |
XM_008763053.1 | XP_008761275.1 | 6 - 7 | 1344 | 113 | ||
GenBank mRNA | AY081195.1 | - | 5 - 6 | 569 | 113 | |
BC107920.1 | - | 6 - 7 | 686 | 113 | ||
FQ227013.1 | - | 6 - 7 | 1312 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
