Assay Details
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037768.1 | NP_001032857.1 | 2 - 3 | 153 | 66 | |
XM_006236740.2 | XP_006236802.1 | 2 - 3 | 1209 | 66 | ||
GenBank mRNA | BC079456.1 | - | 2 - 3 | 135 | 66 | |
BC107647.1 | - | 2 - 3 | 153 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
