Assay Details
Gene Symbol: Htra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106599.2 | NP_001100069.1 | 7 - 8 | 1239 | 110 | |
GenBank mRNA | BC087588.1 | - | 6 - 7 | 1113 | 110 | |
BC158760.1 | - | 7 - 8 | 1239 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
