Assay Details
Gene Symbol: Htra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106599.2 | NP_001100069.1 | 4 - 5 | 964 | 98 | |
GenBank mRNA | BC158760.1 | - | 4 - 5 | 964 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
