Assay Details
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109398.1 | NP_001102868.1 | 1 - 2 | 177 | 101 | |
GenBank mRNA | FQ234580.1 | - | 1 - 2 | 126 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
