Assay Details
Gene Symbol: Il5ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053645.1 | NP_446097.1 | 5 - 6 | 700 | 132 | |
XM_006236977.2 | XP_006237039.1 | 7 - 8 | 1091 | 132 | ||
GenBank mRNA | AB056101.1 | - | 5 - 6 | 700 | 132 | |
AF324153.1 | - | 5 - 6 | 700 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
