Assay Details
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100750.1 | NP_001094220.1 | 10 - 11 | 1192 | 80 | |
GenBank mRNA | BC079367.1 | - | 10 - 11 | 1185 | 80 | |
BC089863.1 | - | 9 - 10 | 1050 | 80 | ||
BC099746.1 | - | 10 - 11 | 1176 | 80 | ||
BC160886.1 | - | 10 - 11 | 1192 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P181503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150937 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P181503 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P181503 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P181503 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P181503 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20118 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71996 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45365 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17514 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88834 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27306 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63213 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3500 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69656 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33771 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10932 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30338 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15459 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10864308 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864310 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864304 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864303 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864306 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864307 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864309 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10864302 | - | - |
Rat Genome 230 2.0 Array | 1373017_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373017_at | - | - |
Affymetrix HT Rat Focus Array | 1373017_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373017_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400024091.rn.1 | TC0400003402.rn.1 | RTA TC |
Rat Expression Set 230 | 1373017_at | - | - |
Rat Expression Set 230 | 1373017_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864302 | - | - |
Rat Genome U34 Set | rc_AA850579_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20782765 | - | - |
