Assay Details
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100750.1 | NP_001094220.1 | 2 - 3 | 234 | 92 | |
GenBank mRNA | BC079367.1 | - | 2 - 3 | 227 | 92 | |
BC089863.1 | - | 1 - 2 | 92 | 92 | ||
BC099746.1 | - | 2 - 3 | 218 | 92 | ||
BC160886.1 | - | 2 - 3 | 234 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
