Assay Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134351.1 | NP_599178.1 | 5 - 6 | 630 | 66 | |
GenBank mRNA | BC062394.1 | - | 663 | 66 | ||
J05571.1 | - | 5 - 6 | 630 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
