Assay Details
Gene Symbol: Eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106612.1 | NP_001100082.1 | 3 - 4 | 310 | 86 | |
XM_008763159.1 | XP_008761381.1 | 3 - 4 | 724 | 86 | ||
XM_008763160.1 | XP_008761382.1 | 5 - 6 | 794 | 86 | ||
XM_008763161.1 | XP_008761383.1 | 4 - 5 | 658 | 86 | ||
GenBank mRNA | BC168936.1 | - | 3 - 4 | 297 | 86 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008763160.1, XM_008763161.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
