Assay Details
Gene Symbol: Hmces | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025047.1 | NP_001020218.1 | 3 - 4 | 485 | 97 | |
XM_006236851.2 | XP_006236913.1 | 4 - 5 | 869 | 97 | ||
GenBank mRNA | BC083690.1 | - | 3 - 4 | 485 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
