Assay Details
Gene Symbol: Gmcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010956.2 | NP_001010956.1 | 5 - 6 | 900 | 108 | |
NM_001033931.1 | NP_001029103.1 | 5 - 6 | 900 | 108 | ||
GenBank mRNA | AY862307.1 | - | 5 - 6 | 727 | 108 | |
AY862308.1 | - | 5 - 6 | 695 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
