Assay Details
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024783.1 | NP_001019954.1 | 2 - 3 | 345 | 81 | |
XM_006237027.2 | XP_006237089.1 | 1 - 2 | 832 | 81 | ||
XM_006237028.2 | XP_006237090.1 | 1 - 2 | 320 | 81 | ||
GenBank mRNA | BC097951.1 | - | 2 - 3 | 345 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006237028.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
