Assay Details
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024783.1 | NP_001019954.1 | 4 - 5 | 539 | 75 | |
XM_006237027.2 | XP_006237089.1 | 3 - 4 | 1026 | 75 | ||
XM_006237028.2 | XP_006237090.1 | 3 - 4 | 584 | 75 | ||
GenBank mRNA | BC097951.1 | - | 4 - 5 | 539 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
