Assay Details
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024783.1 | NP_001019954.1 | 10 - 11 | 1221 | 62 | |
XM_006237027.2 | XP_006237089.1 | 9 - 10 | 1708 | 62 | ||
XM_006237028.2 | XP_006237090.1 | 9 - 10 | 1266 | 62 | ||
GenBank mRNA | BC097951.1 | - | 10 - 11 | 1221 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
