Assay Details
Gene Symbol: Rab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013019.2 | NP_037151.2 | 1 - 2 | 150 | 89 | |
XM_008772600.1 | XP_008770822.1 | 1 - 2 | 201 | 89 | ||
GenBank mRNA | BC062016.1 | - | 1 - 2 | 150 | 89 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC062016.1, NM_013019.2, XM_008772600.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
